accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOBr

CAS RN 30515-28-7

IUPAC Name: 7-bromoheptanoic acid
Synonyms: 7-bromo-heptanoic acid
Molecular Weight (g/mol): 209.08
Molecular Formula: C7H13BrO2
InChi Key: JLPQXFFMVVPIRW-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCCBr
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results