Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Bromoheptanoic Acid 98.0+%, TCI America™
SDP

Catalog No. B36715G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B36715G Supplier TCI America Supplier No. B36715G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 30515-28-7
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.083
MDL Number MFCD00037800
InChI Key JLPQXFFMVVPIRW-UHFFFAOYSA-N
Synonym 7-bromo-heptanoic acid, heptanoic acid, 7-bromo, 7-bromoenanthic acid, omega-bromoheptanoic acid, 7-bromoheptanoicacid, 5-bromopentylacetate;, 7-bromo heptanoic acid, acmc-1aezo, ksc226m8n
PubChem CID 121723
IUPAC Name 7-bromoheptanoic acid
SMILES C(CCCBr)CCC(=O)O

Specifications

Color White-Yellow
Boiling Point 166°C
Quantity 5 g
UN Number 3261
Formula Weight 209.08
Percent Purity ≥98.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material 7-Bromoheptanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.