CAS RN 30525-89-4
Thermo Scientific Chemicals Paraformaldehyde, 4% in PBS
Molecular Formula: CH2O Molecular Weight (g/mol): 30.026 MDL Number: MFCD00133991 InChI Key: WSFSSNUMVMOOMR-UHFFFAOYSA-N Synonym: formalin,methanal,formol,methylene oxide,paraformaldehyde,oxomethane,paraform,formic aldehyde,oxymethylene,methyl aldehyde PubChem CID: 712 ChEBI: CHEBI:16842 IUPAC Name: formaldehyde SMILES: C=O
Paraformaldehyde, 96%, extra pure
CAS: 30525-89-4 Molecular Formula: CH2O Molecular Weight (g/mol): 30.026 InChI Key: WSFSSNUMVMOOMR-UHFFFAOYSA-N Synonym: formalin,methanal,formol,methylene oxide,paraformaldehyde,oxomethane,paraform,formic aldehyde,oxymethylene,methyl aldehyde PubChem CID: 712 ChEBI: CHEBI:16842 IUPAC Name: formaldehyde SMILES: C=O
Paraformaldehyde, 96%
CAS: 30525-89-4 Molecular Formula: CH2O Molecular Weight (g/mol): 30.026 MDL Number: MFCD00133991 InChI Key: WSFSSNUMVMOOMR-UHFFFAOYSA-N Synonym: formalin,methanal,formol,methylene oxide,paraformaldehyde,oxomethane,paraform,formic aldehyde,oxymethylene,methyl aldehyde PubChem CID: 712 ChEBI: CHEBI:16842 IUPAC Name: formaldehyde SMILES: C=O
Paraformaldehyde 90.0+%, TCI America™
CAS: 30525-89-4 Molecular Formula: CH2O Molecular Weight (g/mol): 30.026 MDL Number: MFCD00133991 InChI Key: WSFSSNUMVMOOMR-UHFFFAOYSA-N Synonym: formalin,methanal,formol,methylene oxide,paraformaldehyde,oxomethane,paraform,formic aldehyde,oxymethylene,methyl aldehyde PubChem CID: 712 ChEBI: CHEBI:16842 IUPAC Name: formaldehyde SMILES: C=O
Thermo Scientific Chemicals Paraformaldehyde, 4% in PBS + Mg + EGTA
CAS: 30525-89-4 Molecular Formula: CH2O Molecular Weight (g/mol): 30.026 MDL Number: MFCD00133991 InChI Key: WSFSSNUMVMOOMR-UHFFFAOYSA-N Synonym: formalin,methanal,formol,methylene oxide,paraformaldehyde,oxomethane,paraform,formic aldehyde,oxymethylene,methyl aldehyde PubChem CID: 712 ChEBI: CHEBI:16842 IUPAC Name: formaldehyde SMILES: C=O
Paraformaldehyde MP Biomedicals
CAS: 30525-89-4 Molecular Formula: CH2O Molecular Weight (g/mol): 30.026 InChI Key: WSFSSNUMVMOOMR-UHFFFAOYSA-N Synonym: formalin,methanal,formol,methylene oxide,paraformaldehyde,oxomethane,paraform,formic aldehyde,oxymethylene,methyl aldehyde PubChem CID: 712 ChEBI: CHEBI:16842 IUPAC Name: formaldehyde SMILES: C=O
Paraformaldehyde, 90%, pure
CAS: 30525-89-4 Molecular Formula: CH2O Molecular Weight (g/mol): 30.026 InChI Key: WSFSSNUMVMOOMR-UHFFFAOYSA-N Synonym: formalin,methanal,formol,methylene oxide,paraformaldehyde,oxomethane,paraform,formic aldehyde,oxymethylene,methyl aldehyde PubChem CID: 712 ChEBI: CHEBI:16842 IUPAC Name: formaldehyde SMILES: C=O
Paraformaldehyde Solution, MP Biomedicals™
CAS: 30525-89-4 Molecular Formula: CH2O Molecular Weight (g/mol): 30.03 InChI Key: WSFSSNUMVMOOMR-UHFFFAOYSA-N
Paraformaldehyde, MP Biomedicals™
CAS: 30525-89-4 Molecular Formula: CH2O Molecular Weight (g/mol): 30.026 InChI Key: WSFSSNUMVMOOMR-UHFFFAOYSA-N Synonym: formalin,methanal,formol,methylene oxide,paraformaldehyde,oxomethane,paraform,formic aldehyde,oxymethylene,methyl aldehyde PubChem CID: 712 ChEBI: CHEBI:16842 IUPAC Name: formaldehyde SMILES: C=O
Sigma Aldrich Paraformaldehyde
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Paraformaldehyde
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Formaldehyde, Para (Certified), Fisher Chemical
CAS: 30525-89-4 Molecular Formula: CH2O Molecular Weight (g/mol): 30.026 MDL Number: MFCD00133991 InChI Key: WSFSSNUMVMOOMR-UHFFFAOYSA-N Synonym: formalin,methanal,formol,methylene oxide,paraformaldehyde,oxomethane,paraform,formic aldehyde,oxymethylene,methyl aldehyde PubChem CID: 712 ChEBI: CHEBI:16842 IUPAC Name: formaldehyde SMILES: C=O
Avantor Paraformaldehyde, BAKER™, J.T. Baker™
CAS: 30525-89-4 Molecular Formula: CH2O Molecular Weight (g/mol): 30.026 InChI Key: WSFSSNUMVMOOMR-UHFFFAOYSA-N Synonym: formalin,methanal,formol,methylene oxide,paraformaldehyde,oxomethane,paraform,formic aldehyde,oxymethylene,methyl aldehyde PubChem CID: 712 ChEBI: CHEBI:16842 IUPAC Name: formaldehyde SMILES: C=O