Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Paraformaldehyde Solution, MP Biomedicals™

Catalog No. MP219998380 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
2 x 10 mL
10 x 10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219998380 10 x 10 mL
MP219998320 2 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP219998380 Supplier MP Biomedicals, Inc Supplier No. 0219998380
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Paraformaldehyde Solution, Methanol Free, clear Liquid

  • Recommended for use in the preparation of formalin fixatives for tissues or cells when the samples are to be used in florescence studies.
  • Methanol free Paraformaldehyde is ready to use solution used as a fixative for electron microscopy.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30525-89-4
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N

Specifications

Quantity 10 x 10 mL
Physical Form Liquid
storageNote1 +4°C, store under nitrogen
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.