Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Paraformaldehyde, MP Biomedicals™

Catalog No. MP021501465 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021501465 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP021501465 Supplier MP Biomedicals, Inc Supplier No. 02150146.5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used as a disinfectant and fumigant. Paraformaldehyde, MP Biomedicals is the smallest polyoxymethylene, a polymerization product of formaldehyde. It is also used in the preparation of formalin fixatives for tissues or cells when the samples are to be used

Paraformaldehyde is used as a fixative in electron microscopy and in the preparation of formalin fixatives for tissues or cells when the samples are to be used in florescence studies. It is also used in manufacturing synthetic resins. It has been used in the past for disinfecting sickrooms, clothing, linen and sickroom utensils, and as a fumigant.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30525-89-4
Molecular Formula CH2O
Molecular Weight (g/mol) 30.026
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Melting Point 120–170°C (Lit.)
Density 0.88g/mL at 25°C (Lit.)
Boiling Point Does not boil, gives off formaldehyde gas when heated
Quantity 500 g
Solubility Information Soluble in water (hot water - ∼37% w/v; slowly soluble in cold water). Insoluble in alcohol or ether.
Formula Weight (30.03)n
Physical Form Powder
Recommended Storage 4°C, Store Under Nitrogen.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.