accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2SNH2NFFF(E/Z)

CAS RN 305350-87-2

IUPAC Name: 3-amino-3-[(4-aminophenyl)sulfanyl]-2-[2-(trifluoromethyl)phenyl]prop-2-enenitrile
Synonyms: 3-amino-3-[(4-aminophenyl)sulfanyl]-2-[2-(trifluoromethyl)phenyl]prop-2-enenitrile
Molecular Weight (g/mol): 335.35
Molecular Formula: C16H12F3N3S
InChi Key: JLOXTZFYJNCPIS-UHFFFAOYSA-N
SMILES: NC(SC1=CC=C(N)C=C1)=C(C#N)C1=CC=CC=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)