accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3055-94-5

IUPAC Name: 2-{2-[2-(dodecyloxy)ethoxy]ethoxy}ethan-1-ol
Synonyms: 2-{2-[2-(dodecyloxy)ethoxy]ethoxy}ethanol
Molecular Weight (g/mol): 318.5
Molecular Formula: C18H38O4
InChi Key: FKMHSNTVILORFA-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCOCCOCCOCCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results