accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFClCl

CAS RN 306-83-2

IUPAC Name: 2,2-dichloro-1,1,1-trifluoroethane
Synonyms: 2,2-dichloro-1,1,1-trifluoroethane
Molecular Weight (g/mol): 152.93
Molecular Formula: C2HCl2F3
InChi Key: OHMHBGPWCHTMQE-UHFFFAOYSA-N
SMILES: FC(F)(F)C(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results