accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3

CAS RN 30673-36-0

IUPAC Name: butyl decanoate
Synonyms: butyl decanoate
Molecular Weight (g/mol): 228.38
Molecular Formula: C14H28O2
InChi Key: ZRNCNTSXSYXHOW-UHFFFAOYSA-N
SMILES: CCCCCCCCCC(=O)OCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results