accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNH2SOOONOO

CAS RN 30693-53-9

IUPAC Name: sodium 2-amino-5-nitrobenzene-1-sulfonate
Synonyms: sodium 2-amino-5-nitrobenzenesulfonate
Molecular Weight (g/mol): 240.16
Molecular Formula: C6H5N2NaO5S
InChi Key: VTMRGLXTSDVXAE-UHFFFAOYSA-M
SMILES: [Na+].NC1=CC=C(C=C1S([O-])(=O)=O)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results