Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 2-Amino-5-nitrobenzenesulfonate 98.0+%, TCI America™
SDP

Catalog No. A037525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A037525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A037525G Supplier TCI America Supplier No. A037525G

missing translation for 'validMainframeMsg'

Specifications

CAS 30693-53-9
Color Yellow
Molecular Formula C6H5N2NaO5S
Quantity 25 g
MDL Number MFCD00068347
Synonym 2-Amino-5-nitrobenzenesulfonic Acid Sodium Salt, 4-Nitroaniline-2-sulfonic Acid Sodium Salt, Sodium 4-Nitroaniline-2-sulfonate
InChI Key VTMRGLXTSDVXAE-UHFFFAOYSA-M
SMILES [Na+].NC1=CC=C(C=C1S([O-])(=O)=O)[N+]([O-])=O
IUPAC Name sodium 2-amino-5-nitrobenzene-1-sulfonate
Molecular Weight (g/mol) 240.16
PubChem CID 121742
Formula Weight 240.16
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 2-Amino-5-nitrobenzenesulfonate
Show More Show Less
EINECSNumber (3)-2079
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.