accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFFFFFFFFFFFFFFFFFFFFFFFFFFF

CAS RN 307-62-0

IUPAC Name: triacontafluorotetradecane
Synonyms: triacontafluorotetradecane
Molecular Weight (g/mol): 738.11
Molecular Formula: C14F30
InChi Key: HYZQZWDYBKVIRI-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results