accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 30903-87-8

IUPAC Name: dilithium(1+) oxalate
Synonyms: dilithium oxalate
Molecular Weight (g/mol): 101.9
Molecular Formula: C2Li2O4
InChi Key: YNQRWVCLAIUHHI-UHFFFAOYSA-L
SMILES: [Li+].[Li+].[O-]C(=O)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results