accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 30925-11-2

IUPAC Name: 2-{[(tert-butoxy)carbonyl](methyl)amino}-2-phenylacetic acid
Synonyms: [(tert-butoxycarbonyl)(methyl)amino](phenyl)acetic acid
Molecular Weight (g/mol): 265.31
Molecular Formula: C14H19NO4
InChi Key: COABPHLHHQAKPL-UHFFFAOYNA-N
SMILES: CN(C(C(O)=O)C1=CC=CC=C1)C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)