CAS RN 30989-81-2
IUPAC Name:
3-amino-2-methylprop-2-enal
Synonyms:
3-amino-2-methylprop-2-enal
Molecular Weight (g/mol):
85.11
Molecular Formula:
C4H7NO
InChi Key:
OEZAAXHZEMTBOV-UHFFFAOYSA-N
SMILES:
CC(=CN)C=O