accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 310-71-4

IUPAC Name: 2-chloro-1-ethoxy-1,1,2-trifluoroethane
Synonyms: ethoxyflurane C4H6ClF3O
Molecular Weight (g/mol): 162.54
Molecular Formula: C4H6ClF3O
InChi Key: WOKICPFFJCXEDW-UHFFFAOYNA-N
SMILES: CCOC(F)(F)C(F)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results