Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-1,1,2-trifluoroethyl Ethyl Ether 98.0+%, TCI America™
SDP

Catalog No. C28625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C28625G Supplier TCI America Supplier No. C28625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 310-71-4
Molecular Formula C4H6ClF3O
Molecular Weight (g/mol) 162.536
MDL Number MFCD00018849
InChI Key WOKICPFFJCXEDW-UHFFFAOYSA-N
PubChem CID 136152
ChEBI CHEBI:34266
IUPAC Name 2-chloro-1-ethoxy-1,1,2-trifluoroethane
SMILES CCOC(C(F)Cl)(F)F

Specifications

Color Colorless
Boiling Point 88°C
Quantity 5 g
UN Number 3271
Formula Weight 162.54
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-1,1,2-trifluoroethyl Ethyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.