accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOClTiClClCl

CAS RN 31011-57-1

IUPAC Name: bis(oxolane); tetrachlorotitanium
Synonyms: bis(THF); titanium tetrachloride
Molecular Weight (g/mol): 333.88
Molecular Formula: C8H16Cl4O2Ti
InChi Key: LXWBMENBONGPSB-UHFFFAOYSA-J
SMILES: Cl[Ti](Cl)(Cl)Cl.C1CCOC1.C1CCOC1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)