Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrachlorobis(tetrahydrofuran)titanium(IV), 98%, Thermo Scientific Chemicals

Catalog No. AA3959409
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3959409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3959409 Supplier Thermo Scientific Chemicals Supplier No. 03959409
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 31011-57-1 | Molecular Formula: C8H16Cl4O2Ti | Molecular Weight: 333.881 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31011-57-1
Molecular Formula C8H16Cl4O2Ti
Molecular Weight (g/mol) 333.881
MDL Number MFCD00077884
InChI Key LXWBMENBONGPSB-UHFFFAOYSA-J
Synonym tetrachlorobis tetrahydrofuran titanium, tetrachlorobis tetrahydrofuran titanium iv, bis tetrahydrofuran ; titanium tetrachloride, titanium iv chloride tetrahydrofuran complex
PubChem CID 10958563
IUPAC Name oxolane;tetrachlorotitanium
SMILES C1CCOC1.C1CCOC1.Cl[Ti](Cl)(Cl)Cl

Specifications

Melting Point 126°C to 128°C
Color Yellow
Flash Point -17°C (1°F)
Quantity 10 g
UN Number UN2925
Sensitivity Moisture sensitive
Solubility Information Reacts with water
Formula Weight 333.91
Percent Purity 98%
Physical Form Crystalline or Powder
Chemical Name or Material Tetrachlorobis(tetrahydrofuran)titanium(IV)
Show More Show Less
Hazard Category H228-H314-H335
Hazard Statement GHS H Statement
H228-H314-H318
Flammable solid.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.