CAS RN 3102-57-6
IUPAC Name:
N-[(2S,3R,4E)-1,3-dihydroxyoctadec-4-en-2-yl]acetamide
Synonyms:
C2-ceramide N-acetylsphingosine N-(acetyl)-ceramide
Molecular Weight (g/mol):
341.54
Molecular Formula:
C20H39NO3
InChi Key:
BLTCBVOJNNKFKC-QUDYQQOWSA-N
SMILES:
CCCCCCCCCCCCC\C=C\[C@@H](O)[C@H](CO)NC(C)=O
1
–
2
of
2
results
1
Promotions Available
Thermo Scientific Chemicals N-Acetyl-D-sphingosine, 98%
Thermo Scientific Chemicals N-Acetyl-D-sphingosine, 98%
CAS: 3102-57-6 Molecular Formula: C20H39NO3 Synonym: Acetyl ceramide; C2-Ceramide
2
R&D Systems™ Ceramide
CAS: 3102-57-6 Molecular Formula: C20H39NO3