accessibility menu, dialog, popup

UserName

Viewing profile as user:
MgCH3OHN(S)OOONH(S)OOOHO

CAS RN 3106-85-2

IUPAC Name: magnesium(2+) (4S)-4-carboxy-4-[(2S)-3-carboxylato-2-acetamidopropanamido]butanoate
Synonyms: N-acetylaspartylglutamic acid
Molecular Weight (g/mol): 326.54
Molecular Formula: C11H14MgN2O8
InChi Key: BQAXDBLSAIPBND-LEUCUCNGSA-L
SMILES: [Mg++].CC(=O)N[C@@H](CC([O-])=O)C(=O)N[C@@H](CCC([O-])=O)C(O)=O
Quantity 
  • (1)
  • (1)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results