Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-ASP-GLU, MP Biomedicals™

Catalog No. ICN15303683 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15303683 250 mg
ICN15303680 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15303683 Supplier MP Biomedicals, Inc Supplier No. 0215303683
Neurotransmitter
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3106-85-2
Molecular Formula C11H14MgN2O8
Molecular Weight (g/mol) 326.54
MDL Number MFCD00076652
InChI Key BQAXDBLSAIPBND-LEUCUCNGSA-L
Synonym isospaglumic acid, spaglumic acid, naag, n-acetylaspartylglutamate, n-acetyl-asp-glu, unii-1w8m12wxyl, n-acetylaspartylglutamic acid, 1w8m12wxyl, n-acetyl-l-aspartyl-l-glutamic acid, n-acetyl-l-alpha-aspartyl-l-glutamic acid
PubChem CID 188803
ChEBI CHEBI:73688
IUPAC Name magnesium(2+) (4S)-4-carboxy-4-[(2S)-3-carboxylato-2-acetamidopropanamido]butanoate
SMILES [Mg++].CC(=O)N[C@@H](CC([O-])=O)C(=O)N[C@@H](CCC([O-])=O)C(O)=O

Specifications

Color White
Quantity 250 mg
Formula Weight 304.3
Physical Form Powder
Chemical Name or Material N-Acetyl-ASP-GLU
storageNote1 Store desiccated.
Recommended Storage Store at -20°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.