CAS RN 31161-17-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
CERILLIANT™ EDDP perchlorate, , Sold by MilliporeSigma™ Supelco™
Certified reference material