accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2COOHNHNSOONOO

CAS RN 312283-45-7

IUPAC Name: prop-2-en-1-yl N-[3-(2-nitrobenzenesulfonamido)propyl]carbamate
Synonyms: prop-2-en-1-yl N-[3-(2-nitrobenzenesulfonamido)propyl]carbamate
Molecular Weight (g/mol): 343.35
Molecular Formula: C13H17N3O6S
InChi Key: INKIDQWOEVNHHA-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1S(=O)(=O)NCCCNC(=O)OCC=C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results