accessibility menu, dialog, popup

UserName

Viewing profile as user:
PbPb

CAS RN 3124-01-4

IUPAC Name: hexaphenyldilead
Synonyms: hexaphenyldilead
Molecular Weight (g/mol): 877
Molecular Formula: C36H30Pb2
InChi Key: SOLOHWWGJALNFO-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)[Pb](C1=CC=CC=C1)(C1=CC=CC=C1)[Pb](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results