Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexaphenyldilead, 98%, Thermo Scientific™

Catalog No. AAA1663706
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1663706 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1663706 Supplier Thermo Scientific Chemicals Supplier No. A1663706

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3124-01-4
Molecular Formula C36H30Pb2
Linear Formula (C6H5)6Pb2
Quantity 5g
MDL Number MFCD00014066
UN Number UN3077
Synonym hexaphenyldilead, hexaphenyldiplumbane, dilead, hexaphenyl, lead, hexaphenyldi, diplumbane, hexaphenyl, unii-km5rj5fzy2, km5rj5fzy2, diplumbane, 1,1,1,2,2,2-hexaphenyl, hexaphenyldilead iv
InChI Key SOLOHWWGJALNFO-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)[Pb](C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)[Pb](C2=CC=CC=C2)C3=CC=CC=C3
IUPAC Name triphenyllead
Molecular Weight (g/mol) 877.00
PubChem CID 6327846
Formula Weight 877.02
Percent Purity 98%
Chemical Name or Material Hexaphenyldilead
Show More Show Less
Hazard Category H302+H332-H350-H360Df-H373
Hazard Statement GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 221-505-4
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.