CAS RN 312693-01-9
IUPAC Name:
zinc(2+) 6-ethoxy-6-oxohexan-1-ide bromide
Synonyms:
zinc(2+) 6-ethoxy-6-oxohexan-1-ide bromide
Molecular Weight (g/mol):
288.49
Molecular Formula:
C8H15BrO2Zn
InChi Key:
ZISDBYYELZLQGH-UHFFFAOYSA-M
SMILES:
[Zn++].[Br-].CCOC(=O)CCCC[CH2-]
1
–
1
of
1
results
1
Sigma Aldrich 3-Ethoxy-3-Oxopropylzinc Bromide Solution
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.