CAS RN 313553-47-8
IUPAC Name:
N-[4-(2,4-dimethylphenyl)-1,3-thiazol-2-yl]benzamide
Synonyms:
N-[4-(2,4-dimethylphenyl)-1,3-thiazol-2-yl]benzamide
Molecular Weight (g/mol):
308.4
Molecular Formula:
C18H16N2OS
InChi Key:
JPMOKRWIYQGMJL-UHFFFAOYSA-N
SMILES:
CC1=CC(C)=C(C=C1)C1=CSC(NC(=O)C2=CC=CC=C2)=N1
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Hec1/Nek2 Mitotic Pathway Inhibitor I, INH1
The Hec1/Nek2 Mitotic Pathway Inhibitor I, INH1, also referenced under CAS 313553-47-8, controls the biological activity of Hec1/Nek2 Mitotic Pathway.