accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 31515-43-2

IUPAC Name: 1-(benzenesulfonyl)-2-nitrobenzene
Synonyms: 1-(benzenesulfonyl)-2-nitrobenzene
Molecular Weight (g/mol): 263.27
Molecular Formula: C12H9NO4S
InChi Key: GKNMUCPEXSCGKC-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1S(=O)(=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results