Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitrophenyl Phenyl Sulfone 98.0+%, TCI America™
SDP

Catalog No. N076225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N076225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N076225G Supplier TCI America Supplier No. N076225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 31515-43-2
Molecular Formula C12H9NO4S
Molecular Weight (g/mol) 263.267
InChI Key GKNMUCPEXSCGKC-UHFFFAOYSA-N
Synonym 2-Nitrodiphenyl Sulfone, 1-Nitro-2-(phenylsulfonyl)benzene
PubChem CID 64984
IUPAC Name 1-(benzenesulfonyl)-2-nitrobenzene
SMILES C1=CC=C(C=C1)S(=O)(=O)C2=CC=CC=C2[N+](=O)[O-]

Specifications

Melting Point 147°C
Color White-Yellow
Quantity 25 g
Formula Weight 263.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitrophenyl Phenyl Sulfone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.