accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2CNHHNONNO(E/Z)

CAS RN 315183-21-2

IUPAC Name: 2-(4-benzylpiperazin-1-yl)-N'-{[6-oxo-5-(prop-2-en-1-yl)cyclohexa-2,4-dien-1-ylidene]methyl}acetohydrazide
Synonyms: pac-1
Molecular Weight (g/mol): 392.5
Molecular Formula: C23H28N4O2
InChi Key: XSGXRYNZNGUGEJ-UHFFFAOYSA-N
SMILES: C=CCC1=CC=CC(=CNNC(=O)CN2CCN(CC3=CC=CC=C3)CC2)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results