accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOH3CH3CCH3CH3VO

CAS RN 3153-26-2

IUPAC Name: oxovanadiumbis(ylium) bis((2Z)-4-oxopent-2-en-2-olate)
Synonyms: oxovanadium(IV) bis((2Z)-4-oxopent-2-en-2-olate)
Molecular Weight (g/mol): 265.16
Molecular Formula: C10H14O5V
InChi Key: JFHJZWAQYMGNBE-SUKNRPLKSA-L
SMILES: O=[V++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
Percent Purity 
  • (1)
  • (1)
  • (2)
  • (3)
Physical Form 
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (6)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (3)
  • (4)
  • (3)