accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClFONHHNF

CAS RN 31540-62-2

IUPAC Name: 8-fluoro-2-[4-(4-fluorophenyl)-4-oxobutyl]-1H,2H,3H,4H,5H-pyrido[4,3-b]indol-2-ium chloride
Synonyms: 8-fluoro-2-[4-(4-fluorophenyl)-4-oxobutyl]-1H,2H,3H,4H,5H-pyrido[4,3-b]indol-2-ium chloride
Molecular Weight (g/mol): 390.86
Molecular Formula: C21H21ClF2N2O
InChi Key: LGNFRLBPCVLGOB-UHFFFAOYNA-N
SMILES: [Cl-].FC1=CC=C(C=C1)C(=O)CCC[NH+]1CCC2=C(C1)C1=CC(F)=CC=C1N2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results