accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOH

CAS RN 31554-15-1

IUPAC Name: [3-(hydroxymethyl)naphthalen-2-yl]methanol
Synonyms: [3-(hydroxymethyl)naphthalen-2-yl]methanol
Molecular Weight (g/mol): 188.23
Molecular Formula: C12H12O2
InChi Key: DTYJRRQQOLBRGH-UHFFFAOYSA-N
SMILES: OCC1=CC2=CC=CC=C2C=C1CO
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results