accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SSNH3CFFFFOHOO

CAS RN 317318-84-6

IUPAC Name: 2-{4-[({2-[3-fluoro-4-(trifluoromethyl)phenyl]-4-methyl-1,3-thiazol-5-yl}methyl)sulfanyl]-2-methylphenoxy}acetic acid
Synonyms: 4-[({2-[3-fluoro-4-(trifluoromethyl)phenyl]-4-methyl-1,3-thiazol-5-yl}methyl)sulfanyl]-2-methylphenoxyacetic acid
Molecular Weight (g/mol): 471.48
Molecular Formula: C21H17F4NO3S2
InChi Key: HWVNEWGKWRGSRK-UHFFFAOYSA-N
SMILES: CC1=C(CSC2=CC=C(OCC(O)=O)C(C)=C2)SC(=N1)C1=CC=C(C(F)=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results