CAS RN 3179-63-3
IUPAC Name:
3-(dimethylamino)propan-1-ol
Synonyms:
1-propanol, 3-(dimethylamino)-
Molecular Weight (g/mol):
103.17
Molecular Formula:
C5H13NO
InChi Key:
PYSGFFTXMUWEOT-UHFFFAOYSA-N
SMILES:
CN(C)CCCO
1
–
2
of
2
results
2
Sigma Aldrich 3-Dimethylamino-1-propanol
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.