accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 318233-97-5

IUPAC Name: 2,4-dimethyl 1,3-thiazolidine-2,4-dicarboxylate
Synonyms: 2,4-dimethyl 1,3-thiazolidine-2,4-dicarboxylate
Molecular Weight (g/mol): 205.23
Molecular Formula: C7H11NO4S
InChi Key: ZZAHPKXSIYXTAP-UHFFFAOYNA-N
SMILES: COC(=O)C1CSC(N1)C(=O)OC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results