accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH2H2COOH3C

CAS RN 3195-24-2

IUPAC Name: 1,3-diethyl 2,2-bis(prop-2-en-1-yl)propanedioate
Synonyms: 1,3-diethyl 2,2-bis(prop-2-en-1-yl)propanedioate
Molecular Weight (g/mol): 240.3
Molecular Formula: C13H20O4
InChi Key: LYUUVYQGUMRKOV-UHFFFAOYSA-N
SMILES: CCOC(=O)C(CC=C)(CC=C)C(=O)OCC
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (1)
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (1)
  • (3)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (4)
  • (5)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)