Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl 2,2-diallylmalonate, Tech., Thermo Scientific™

Catalog No. SB00826EE
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB00826EE 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SB00826EE Supplier Thermo Scientific Chemicals Supplier No. SB00826EE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3195-24-2
Molecular Formula C13H20O4
Molecular Weight (g/mol) 240.299
MDL Number MFCD00009126
InChI Key LYUUVYQGUMRKOV-UHFFFAOYSA-N
Synonym diethyl diallylmalonate, diethyl 2,2-diallylmalonate, diallylmalonic acid diethyl ester, propanedioic acid, di-2-propenyl-, diethyl ester, 1,3-diethyl 2,2-bis prop-2-en-1-yl propanedioate, diethyl 2,2-diprop-2-enylpropane-1,3-dioate, diethyldiallylmalonate, malonic acid, diallyl-, diethyl ester, diethyl2,2-diallylmalonate, acmc-1cp69
PubChem CID 76664
IUPAC Name diethyl 2,2-bis(prop-2-enyl)propanedioate
SMILES CCOC(=O)C(CC=C)(CC=C)C(=O)OCC

Specifications

Quantity 50 g
Formula Weight 240.3
Percent Purity Tech.
Grade Technical
Chemical Name or Material Diethyl 2, 2-diallylmalonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.