accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSHOOFH3C(E)

CAS RN 32004-67-4

IUPAC Name: 2-[(1E)-5-fluoro-2-methyl-1-{[4-(methylsulfanyl)phenyl]methylidene}-1H-inden-3-yl]acetic acid
Synonyms: [(3E)-6-fluoro-2-methyl-3-{[4-(methylsulfanyl)phenyl]methylidene}inden-1-yl]acetic acid
Molecular Weight (g/mol): 340.41
Molecular Formula: C20H17FO2S
InChi Key: LFWHFZJPXXOYNR-RQZCQDPDSA-N
SMILES: CSC1=CC=C(\C=C2/C(C)=C(CC(O)=O)C3=CC(F)=CC=C23)C=C1
Product Type 
  • (1)
  • (1)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results