accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32072-96-1

IUPAC Name: 3-[(1E)-hexadec-1-en-1-yl]oxolane-2,5-dione
Synonyms: 3-[(1E)-hexadec-1-en-1-yl]oxolane-2,5-dione
Molecular Weight (g/mol): 322.49
Molecular Formula: C20H34O3
InChi Key: RSPWVGZWUBNLQU-FOCLMDBBNA-N
SMILES: CCCCCCCCCCCCCC\C=C\C1CC(=O)OC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results