accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONNClN

CAS RN 321432-82-0

IUPAC Name: 4-chloro-5-methoxy-2-(pyridin-2-yl)pyrimidine
Synonyms: 4-chloro-5-methoxy-2-(pyridin-2-yl)pyrimidine
Molecular Weight (g/mol): 221.64
Molecular Formula: C10H8ClN3O
InChi Key: BTHRCJACYHBCHX-UHFFFAOYSA-N
SMILES: COC1=CN=C(N=C1Cl)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results