accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 321724-19-0

IUPAC Name: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine
Synonyms: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine
Molecular Weight (g/mol): 206.05
Molecular Formula: C10H15BN2O2
InChi Key: WSRGGSAGSUEJKQ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CN=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results