Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

pyrimidine-5-boronic acid, 97%, Thermo Scientific™

Catalog No. MAY00076DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MAY00076DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAY00076DA Supplier Thermo Scientific Chemicals Supplier No. MAY00076DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 321724-19-0
Molecular Formula C10H15BN2O2
Molecular Weight (g/mol) 206.05
MDL Number MFCD05155223
InChI Key WSRGGSAGSUEJKQ-UHFFFAOYSA-N
Synonym 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, pyrimidine-5-boronic acid pinacol ester, pinacol ester pyrimidinyl-5-boronic acid, 5-pyrimidineboronic acid pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, pyrimidyl-5-boronic acid pinacolate, pyrimidine-5-boronic acid, pinacol ester, pyrimidine, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, pyrimidin-5-ylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-pyrimidin-5-yl-1,3,2-dioxaborolane
PubChem CID 4192667
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine
SMILES CC1(C)OB(OC1(C)C)C1=CN=CN=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 206.05
Percent Purity 97%
Chemical Name or Material Pyrimidine-5-boronic acid, pinacol ester
Hazard Category Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.