accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(R)H3CH3COH(R)(S)(R)CH3(R)OH(S)HOH2C(E)(Z)

CAS RN 32222-06-3

IUPAC Name: (1R,3S,5Z)-5-{2-[(1R,3aS,4E,7aR)-1-[(2R)-6-hydroxy-6-methylheptan-2-yl]-7a-methyl-octahydro-1H-inden-4-ylidene]ethylidene}-4-methylidenecyclohexane-1,3-diol
Synonyms: calcitriol calcitriol galderma brand 1,25(oh)>2<d>3< preparation D3, 1,25-dihydroxyvitamin kyramed, calcitriol 1,25 dihydroxyvitamin d3 1,25-dihydroxyvitamin D calcijex alphapharm brand of calcitriol 1α,25-dihydroxyvitamin D
Molecular Weight (g/mol): 416.65
Molecular Formula: C27H44O3
InChi Key: GMRQFYUYWCNGIN-NKMMMXOESA-N
SMILES: C[C@H](CCCC(C)(C)O)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1\C[C@@H](O)C[C@H](O)C1=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)