accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CNOH

CAS RN 322473-89-2

IUPAC Name: 8-[bis(2-methylphenyl)methyl]-3-phenyl-8-azabicyclo[3.2.1]octan-3-ol
Synonyms: 8-[bis(2-methylphenyl)methyl]-3-phenyl-8-azabicyclo[3.2.1]octan-3-ol
Molecular Weight (g/mol): 397.56
Molecular Formula: C28H31NO
InChi Key: LOSJNRBXNQTUNT-UHFFFAOYNA-N
SMILES: CC1=CC=CC=C1C(N1C2CCC1CC(O)(C2)C1=CC=CC=C1)C1=CC=CC=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results