accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OPPO

CAS RN 32305-98-9

IUPAC Name: ({5-[(diphenylphosphanyl)methyl]-2,2-dimethyl-1,3-dioxolan-4-yl}methyl)diphenylphosphane
Synonyms: DIOP
Molecular Weight (g/mol): 498.54
Molecular Formula: C31H32O2P2
InChi Key: VCHDBLPQYJAQSQ-UHFFFAOYNA-N
SMILES: CC1(C)OC(CP(C2=CC=CC=C2)C2=CC=CC=C2)C(CP(C2=CC=CC=C2)C2=CC=CC=C2)O1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)