accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHONHHNO(E)

CAS RN 3232-36-8

IUPAC Name: 2-hydroxy-N'-{[(1E)-6-oxocyclohexa-2,4-dien-1-ylidene]methyl}benzohydrazide
Synonyms: 2-hydroxy-N'-{[(1E)-6-oxocyclohexa-2,4-dien-1-ylidene]methyl}benzohydrazide
Molecular Weight (g/mol): 256.26
Molecular Formula: C14H12N2O3
InChi Key: NCCJLHWEUIAECU-MDZDMXLPSA-N
SMILES: OC1=CC=CC=C1C(=O)NN\C=C1/C=CC=CC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results