accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32345-60-1

IUPAC Name: methyl 2-(2-chlorophenyl)-2-hydroxyacetate
Synonyms: methyl 2-(2-chlorophenyl)-2-hydroxyacetate
Molecular Weight (g/mol): 200.62
Molecular Formula: C9H9ClO3
InChi Key: ZMPGBVQQIQSQED-UHFFFAOYNA-N
SMILES: COC(=O)C(O)C1=CC=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results