accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH3CHBr

CAS RN 32353-50-7

IUPAC Name: 1-ethyl-2-methylpyridin-1-ium hydrobromide
Synonyms: 1-ethyl-2-methylpyridin-1-ium hydrobromide
Molecular Weight (g/mol): 203.1
Molecular Formula: C8H13BrN
InChi Key: QTXZDFXPUPKBJC-UHFFFAOYSA-N
SMILES: Br.CC[N+]1=CC=CC=C1C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results